siRNA Details
Gene Symbol: Tox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199392.2 | NP_955424.2 | 3 | 309 | ||
XM_006235578.2 | XP_006235640.1 | 3 | 343 | |||
GenBank mRNA | AB096685.1 | - | 3 | 310 |

Gene Symbol: Tox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199392.2 | NP_955424.2 | 3 | 309 | ||
XM_006235578.2 | XP_006235640.1 | 3 | 343 | |||
GenBank mRNA | AB096685.1 | - | 3 | 310 |