Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Aqp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142366.2 | NP_001135838.1 | 2 | 237 | ||
NM_001270558.2 | NP_001257487.1 | 2 | 229 | |||
NM_001270559.2 | NP_001257488.1 | 2 | 237 | |||
NM_012825.4 | NP_036957.1 | 2 | 229 | |||
GenBank mRNA | L27588.1 | - | 1 | 97 | ||
U14007.1 | - | 2 | 163 |