Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033665.1 | NP_001028837.1 | 5, 6 | 1022 | ||
NM_012916.2 | NP_037048.2 | 5, 6 | 1022 | |||
XM_006232595.2 | XP_006232657.1 | 5, 6 | 2103 | |||
XM_006232596.2 | XP_006232658.1 | 5, 6 | 1006 | |||
XM_006232597.2 | XP_006232659.1 | 5, 6 | 824 | |||
GenBank mRNA | BC094307.1 | - | 1, 2 | 1802 | ||
U37142.1 | - | 5, 6 | 826 | |||
X79881.1 | - | 5, 6 | 876 | |||
X86406.1 | - | 5, 6 | 876 | |||
Z28366.1 | - | 5, 6 | 1017 |