Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NT5C2 SIRNA Assays ›
Gene Symbol:
NT5C2
Gene Name:
5'-nucleotidase, cytosolic II
Gene Aliases:
GMP, NT5B, PNT5, SPG45, SPG65, cN-II
Chromosome Location:
Chr.10: 103088017 - 103193306 on Build GRCh38
UniGene ID:
Hs.97439
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
19745
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NT5C2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134373.2
NP_001127845.1
8
700
NM_012229.4
NP_036361.1
9
798
XM_005269632.4
XP_005269689.1
9
709
XM_005269633.4
XP_005269690.1
10
802
XM_005269634.4
XP_005269691.1
10
834
XM_005269635.4
XP_005269692.1
9
1024
XM_005269636.4
XP_005269693.1
9
810
XM_005269637.4
XP_005269694.1
8
1528
XM_005269638.4
XP_005269695.1
8
2829
XM_005269639.4
XP_005269696.1
7
553
XM_005269640.4
XP_005269697.1
10
825
XM_005269641.4
XP_005269698.1
9
727
XM_005269642.4
XP_005269699.1
8
652
XM_005269643.4
XP_005269700.1
7
592
XM_005269644.4
XP_005269701.1
11
849
XM_005269645.4
XP_005269702.1
10
751
XM_005269646.4
XP_005269703.1
9
677
XM_006717721.3
XP_006717784.1
9
751
XM_006717723.3
XP_006717786.1
7
548
XM_011539537.2
XP_011537839.1
10
834
XM_017015947.1
XP_016871436.1
7
578
XM_017015948.1
XP_016871437.1
10
934
XM_017015949.1
XP_016871438.1
10
919
XM_017015950.1
XP_016871439.1
8
635
XM_017015951.1
XP_016871440.1
7
561
XM_017015952.1
XP_016871441.1
9
858
XM_017015953.1
XP_016871442.1
10
1253
XM_017015954.1
XP_016871443.1
9
1192
XM_017015955.1
XP_016871444.1
9
879
XM_017015956.1
XP_016871445.1
9
1085
XM_017015957.1
XP_016871446.1
11
943
XM_017015958.1
XP_016871447.1
12
1062
XM_017015959.1
XP_016871448.1
8
653
XM_017015960.1
XP_016871449.1
9
616
XM_017015961.1
XP_016871450.1
9
1011
XM_017015962.1
XP_016871451.1
9
845
XM_017015963.1
XP_016871452.1
9
949
XM_017015964.1
XP_016871453.1
9
928
XM_017015965.1
XP_016871454.1
8
805
XM_017015966.1
XP_016871455.1
8
805
XM_017015967.1
XP_016871456.1
8
740
XM_017015968.1
XP_016871457.1
9
733
XM_017015969.1
XP_016871458.1
9
827
XM_017015970.1
XP_016871459.1
7
1470
XM_017015971.1
XP_016871460.1
6
514
XM_017015972.1
XP_016871461.1
9
727
XM_017015973.1
XP_016871462.1
10
751
XM_017015974.1
XP_016871463.1
9
751
XM_017015975.1
XP_016871464.1
7
592
XM_017015976.1
XP_016871465.1
7
598
GenBank mRNA
AK127670.1
-
9
789
AK291667.1
-
8
666
AK295593.1
-
7
842
AK312517.1
-
8
666
BC001595.1
-
8
749
3543103
BX648078.1
-
11
1044
D38524.1
-
8
688
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
purine nucleotide catabolic process
phosphorylation
dephosphorylation
drug metabolic process
IMP metabolic process
adenosine metabolic process
nucleotide binding
protein binding
5'-nucleotidase activity
metal ion binding
nucleoside phosphotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0