Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242827.1 | NP_001229756.1 | 5, 6 | 720 | ||
NM_012241.4 | NP_036373.1 | 6, 7 | 876 | |||
NM_031244.3 | NP_112534.1 | 6, 7 | 854 | |||
XM_005248968.4 | XP_005249025.1 | 6, 7 | 791 | |||
XM_011514444.2 | XP_011512746.1 | 7, 8 | 925 | |||
XM_017010620.1 | XP_016866109.1 | 5, 6 | 754 | |||
XM_017010621.1 | XP_016866110.1 | 6, 7 | 791 | |||
XM_017010622.1 | XP_016866111.1 | 7, 8 | 984 | |||
XM_017010623.1 | XP_016866112.1 | 7, 8 | 1114 | |||
XM_017010624.1 | XP_016866113.1 | 5, 6 | 754 | |||
XM_017010625.1 | XP_016866114.1 | 5, 6 | 754 | |||
GenBank mRNA | AF083110.1 | - | 6, 7 | 837 | ||
AJ420496.1 | - | 6, 7 | 783 | |||
AK000355.1 | - | 6, 7 | 782 | |||
AK294162.1 | - | 5, 6 | 648 | |||
BC000126.1 | - | 6, 7 | 860 | 3352592 | ||
BX538030.1 | - | 5, 6 | 667 |