siRNA Details
Gene Symbol: Prph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013021.1 | NP_037153.1 | 672 | |||
GenBank mRNA | X52376.1 | - | 672 |

More Information
Panther Classification:
Molecular Function -




