Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013023.2 | NP_037155.2 | 6 | 546 | ||
XM_008767196.1 | XP_008765418.1 | 6 | 809 | |||
GenBank mRNA | M60737.1 | - | 5 | 437 | ||
X15353.1 | - | 5 | 437 | |||
X51781.1 | - | 6 | 546 |