Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013078.1 | NP_037210.1 | 2 | 239 | ||
GenBank mRNA | BC088158.1 | - | 2 | 205 | 7325875 | |
FQ211098.1 | - | 2 | 223 | |||
FQ219225.1 | - | 2 | 226 | |||
FQ219657.1 | - | 2 | 202 | |||
K00001.1 | - | 2 | 225 | |||
K03040.1 | - | 2 | 197 | |||
M11266.1 | - | 2 | 239 | |||
X01178.1 | - | 2 | 239 | |||
X01976.1 | - | 2 | 225 |