Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017141.2 | NP_058837.2 | 6 | 359 | ||
GenBank mRNA | BC098668.1 | - | 6 | 359 | 7386894 | |
J02776.1 | - | 6 | 347 | |||
M13961.1 | - | 6 | 337 | |||
U38801.1 | - | 354 |