siRNA Details
Gene Symbol: Phlda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017180.2 | NP_058876.2 | 1 | 676 | ||
GenBank mRNA | AF192802.1 | - | 1 | 654 |

Gene Symbol: Phlda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017180.2 | NP_058876.2 | 1 | 676 | ||
GenBank mRNA | AF192802.1 | - | 1 | 654 |