siRNA Details
Gene Symbol: SIRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193286.1 | NP_001180215.1 | 13 | 1521 | ||
NM_012237.3 | NP_036369.2 | 16 | 1681 | |||
NM_030593.2 | NP_085096.1 | 15 | 1673 | |||
NR_034146.1 | 14 | 1604 | ||||
XM_006723111.1 | XP_006723174.1 | 17 | 1629 | |||
XM_011526654.1 | XP_011524956.1 | 15 | 1566 | |||
XM_011526655.1 | XP_011524957.1 | 13 | 1381 | |||
XM_017026500.1 | XP_016881989.1 | 15 | 1605 | |||
GenBank mRNA | AB009274.1 | - | 15 | 1398 | ||
AF083107.2 | - | 16 | 1579 | |||
AF095714.1 | - | 15 | 1452 | |||
AF131800.1 | - | 12 | 1176 | |||
AF160214.1 | - | 15 | 1432 | |||
AJ505014.1 | - | 13 | 2151 | |||
AK025876.1 | - | 15 | 1459 | |||
AK054642.1 | - | 16 | 1512 | |||
AK131390.1 | - | 5 | 4346 | |||
AK225824.1 | - | 15 | 1455 | |||
AY030277.1 | - | 15 | 1434 | |||
BC003012.1 | - | 15 | 1524 | 2820929 | ||
BC003547.1 | - | 15 | 1524 | 3536728 | ||
BF346568.1 | - | 1 | 261 |

More Information
Panther Classification:
Molecular Function -





