Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017264.3 | NP_058960.2 | 8, 9 | 631 | ||
GenBank mRNA | BC060574.1 | - | 8, 9 | 635 | 6890794 | |
D45249.1 | - | 8, 9 | 555 | |||
FQ216423.1 | - | 8, 9 | 632 | |||
FQ220175.1 | - | 8, 9 | 580 | |||
FQ224986.1 | - | 8, 9 | 578 | |||
FQ226855.1 | - | 8, 9 | 584 | |||
FQ228562.1 | - | 586 | ||||
FQ230622.1 | - | 8, 9 | 578 | |||
FQ233032.1 | - | 8, 9 | 584 | |||
FQ233635.1 | - | 8, 9 | 584 | |||
FQ233859.1 | - | 8, 9 | 580 |