Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other B2M SIRNA Assays ›
          Gene Symbol:
          B2m
          Gene Name:
          beta-2 microglobulin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 114087287 - 114093311 on Build Rnor_6.0
          UniGene ID:
          Rn.1868
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 199798

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: B2m
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012512.2 NP_036644.1 2 145
          GenBank mRNA FM032790.1 - 2 145
          FQ210051.1 - 2 158
          FQ210252.1 - 2 159
          FQ210804.1 - 2 157
          FQ210906.1 - 2 158
          FQ211296.1 - 2 158
          FQ211419.1 - 2 157
          FQ212410.1 - 2 159
          FQ217926.1 - 2 156
          FQ218141.1 - 2 158
          FQ218251.1 - 2 158
          FQ218298.1 - 2 157
          FQ219157.1 - 2 160
          FQ219455.1 - 707
          FQ220256.1 - 2 154
          FQ221268.1 - 2 158
          FQ221985.1 - 2 157
          FQ222352.1 - 2 155
          FQ225010.1 - 2 158
          FQ225051.1 - 2 162
          FQ225087.1 - 2 157
          FQ225088.1 - 2 159
          FQ225121.1 - 2 157
          FQ225141.1 - 2 157
          FQ225219.1 - 2 156
          FQ225292.1 - 2 158
          FQ225550.1 - 2 157
          FQ225647.1 - 2 157
          FQ225686.1 - 2 160
          FQ225690.1 - 2 299
          FQ225847.1 - 2 157
          FQ225912.1 - 2 157
          FQ226004.1 - 2 158
          FQ226258.1 - 2 160
          FQ226642.1 - 2 159
          FQ226723.1 - 2 157
          FQ227149.1 - 2 157
          FQ227291.1 - 2 163
          FQ227327.1 - 2 158
          FQ227454.1 - 2 158
          FQ227468.1 - 2 156
          FQ227656.1 - 2 158
          FQ227700.1 - 2 158
          FQ227950.1 - 2 156
          FQ227964.1 - 2 157
          FQ227995.1 - 2 153
          FQ228065.1 - 2 159
          FQ228188.1 - 2 158
          FQ228427.1 - 2 159
          FQ229482.1 - 2 150
          FQ230202.1 - 2 158
          FQ230402.1 - 2 158
          FQ230977.1 - 2 157
          FQ231135.1 - 2 158
          FQ231182.1 - 2 158
          FQ231367.1 - 2 157
          FQ231378.1 - 2 157
          FQ231524.1 - 2 157
          FQ231602.1 - 2 158
          FQ231718.1 - 2 157
          FQ231856.1 - 2 157
          FQ231894.1 - 2 157
          FQ232233.1 - 2 158
          FQ232294.1 - 2 158
          FQ232460.1 - 2 157
          FQ232758.1 - 2 157
          FQ232818.1 - 2 157
          FQ232819.1 - 2 157
          FQ232937.1 - 2 157
          FQ232943.1 - 2 159
          FQ232957.1 - 2 155
          FQ233220.1 - 2 158
          FQ233240.1 - 2 158
          FQ233290.1 - 2 157
          FQ233447.1 - 2 158
          FQ233505.1 - 2 157
          FQ233544.1 - 2 157
          FQ233567.1 - 2 162
          FQ233757.1 - 2 157
          FQ233797.1 - 2 156
          FQ233849.1 - 2 157
          FQ234348.1 - 2 158
          FQ234349.1 - 2 157
          FQ234524.1 - 2 155
          FQ234581.1 - 2 155
          FQ234840.1 - 2 157
          FQ234885.1 - 2 157
          FQ235105.1 - 2 157
          FQ235149.1 - 2 155
          FQ235229.1 - 2 157
          Y00441.1 - 2 132
          Y08531.1 - 2 137

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          positive regulation of T cell mediated cytotoxicity
          response to molecule of bacterial origin
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
          T cell differentiation in thymus
          protein refolding
          response to drug
          response to cadmium ion
          iron ion homeostasis
          molecular_function
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0