Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270852.1 | NP_001257781.1 | 14 | 1301 | ||
NM_001270853.1 | NP_001257782.1 | 13 | 1195 | |||
NM_012545.4 | NP_036677.1 | 14 | 1341 | |||
XM_006251474.2 | XP_006251536.1 | 14 | 2077 | |||
XM_006251475.2 | XP_006251537.1 | 14 | 1917 | |||
XM_008770251.1 | XP_008768473.1 | 14 | 1372 | |||
XM_008770252.1 | XP_008768474.1 | 14 | 1435 | |||
XM_008770253.1 | XP_008768475.1 | 13 | 1252 | |||
GenBank mRNA | BC087032.1 | - | 14 | 1301 | 7098083 | |
FQ219697.1 | - | 14 | 1339 | |||
JF273828.1 | - | 13 | 1195 | |||
M27716.1 | - | 14 | 1346 | |||
U31884.1 | - | 14 | 1323 |