Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DRD2 SIRNA Assays ›
          Gene Symbol:
          Drd2
          Gene Name:
          dopamine receptor D2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 53678777 - 53743643 on Build Rnor_6.0
          UniGene ID:
          Rn.87299
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 199873

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Drd2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012547.1 NP_036679.1 5 1045
          XM_006242979.2 XP_006243041.1 5 826
          XM_008766190.1 XP_008764412.1 4 1062
          XM_008766191.1 XP_008764413.1 4 1062
          GenBank mRNA M36831.1 - 5 826
          X53278.1 - 5 825
          X56065.1 - 5 1045

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_010077, BC105666, X55674, BC105665, BC105663, and others.

          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          response to hypoxia
          synaptic transmission, dopaminergic
          startle response
          response to amphetamine
          neurological system process involved in regulation of systemic arterial blood pressure
          regulation of heart rate
          regulation of sodium ion transport
          G-protein coupled receptor internalization
          positive regulation of neuroblast proliferation
          positive regulation of receptor internalization
          G-protein coupled receptor signaling pathway
          adenylate cyclase-modulating G-protein coupled receptor signaling pathway
          negative regulation of adenylate cyclase activity
          adenylate cyclase-inhibiting dopamine receptor signaling pathway
          dopamine receptor signaling pathway
          chemical synaptic transmission
          neuron-neuron synaptic transmission
          axonogenesis
          synapse assembly
          sensory perception of smell
          long-term memory
          grooming behavior
          locomotory behavior
          adult walking behavior
          feeding behavior
          protein localization
          negative regulation of cell proliferation
          associative learning
          visual learning
          response to light stimulus
          response to toxic substance
          response to iron ion
          regulation of dopamine secretion
          response to inactivity
          Wnt signaling pathway
          striatum development
          orbitofrontal cortex development
          cerebral cortex GABAergic interneuron migration
          adenohypophysis development
          cellular potassium ion homeostasis
          negative regulation of cell migration
          peristalsis
          adult behavior
          regulation of cAMP metabolic process
          forebrain development
          auditory behavior
          activation of protein kinase activity
          regulation of synaptic transmission, GABAergic
          positive regulation of cytokinesis
          circadian regulation of gene expression
          negative regulation of dopamine secretion
          response to histamine
          response to nicotine
          intracellular signal transduction
          positive regulation of urine volume
          positive regulation of renal sodium excretion
          positive regulation of multicellular organism growth
          response to cocaine
          negative regulation of circadian sleep/wake cycle, sleep
          dopamine metabolic process
          response to drug
          regulation of potassium ion transport
          response to morphine
          regulation of MAPK cascade
          pigmentation
          regulation of phosphoprotein phosphatase activity
          response to ethanol
          positive regulation of G-protein coupled receptor protein signaling pathway
          negative regulation of blood pressure
          positive regulation of mitotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          phosphatidylinositol metabolic process
          negative regulation of insulin secretion
          acid secretion
          behavioral response to cocaine
          behavioral response to ethanol
          regulation of long-term neuronal synaptic plasticity
          response to axon injury
          branching morphogenesis of a nerve
          arachidonic acid secretion
          negative regulation of protein secretion
          modulation of synaptic transmission
          release of sequestered calcium ion into cytosol
          negative regulation of cytosolic calcium ion concentration
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          regulation of dopamine uptake involved in synaptic transmission
          positive regulation of dopamine uptake involved in synaptic transmission
          regulation of synapse structural plasticity
          negative regulation of protein kinase B signaling
          negative regulation of synaptic transmission, glutamatergic
          positive regulation of growth hormone secretion
          prepulse inhibition
          phospholipase C-activating dopamine receptor signaling pathway
          negative regulation of dopamine receptor signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          regulation of locomotion involved in locomotory behavior
          positive regulation of long-term synaptic potentiation
          negative regulation of voltage-gated calcium channel activity
          dopamine neurotransmitter receptor activity, coupled via Gi/Go
          G-protein coupled receptor activity
          dopamine neurotransmitter receptor activity
          receptor binding
          protein binding
          drug binding
          dopamine binding
          ionotropic glutamate receptor binding
          identical protein binding
          protein homodimerization activity
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0