Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pklr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012624.3 | NP_036756.3 | 8 | 1179 | ||
XM_006232593.2 | XP_006232655.1 | 8 | 1310 | |||
GenBank mRNA | BC161827.1 | - | 7 | 1169 | 7127842 | |
FQ211172.1 | - | 2 | 607 | |||
M11709.1 | - | 8 | 1180 |