Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ESR1 SIRNA Assays ›
          Gene Symbol:
          Esr1
          Gene Name:
          estrogen receptor 1
          Gene Aliases:
          ER-alpha, Esr, RNESTROR
          Chromosome Location:
          Chr.1: 41192150 - 41594799 on Build Rnor_6.0
          UniGene ID:
          Rn.10595
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 200210

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Esr1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012689.1 NP_036821.1 3 772
          XM_006227832.2 XP_006227894.1 3 1372
          XM_008758703.1 XP_008756925.1 4 1420
          XM_008758704.1 XP_008756926.1 3 1344
          GenBank mRNA AB477039.1 - 3 778
          KJ746832.1 - 2 563
          KJ746833.1 - 2 563
          X61098.1 - 2 636
          Y00102.1 - 3 772

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          antral ovarian follicle growth
          epithelial cell development
          osteoblast development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          androgen metabolic process
          male gonad development
          negative regulation of gene expression
          positive regulation of phospholipase C activity
          intracellular steroid hormone receptor signaling pathway
          intracellular estrogen receptor signaling pathway
          intracellular receptor signaling pathway
          response to estradiol
          regulation of apoptotic process
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          steroid hormone mediated signaling pathway
          negative regulation of sequence-specific DNA binding transcription factor activity
          regulation of neuron apoptotic process
          response to estrogen
          positive regulation of nitric oxide biosynthetic process
          positive regulation of epidermal growth factor receptor signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of glucose import
          decidualization
          positive regulation of fibroblast proliferation
          positive regulation of retinoic acid receptor signaling pathway
          negative regulation of smooth muscle cell proliferation
          positive regulation of nitric-oxide synthase activity
          positive regulation of sequence-specific DNA binding transcription factor activity
          Sertoli cell development
          Sertoli cell proliferation
          uterus development
          vagina development
          prostate epithelial cord elongation
          prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
          regulation of branching involved in prostate gland morphogenesis
          mammary gland branching involved in pregnancy
          mammary gland alveolus development
          epithelial cell proliferation involved in mammary gland duct elongation
          positive regulation of ERK1 and ERK2 cascade
          cellular response to estrogen stimulus
          cellular response to estradiol stimulus
          negative regulation of triglyceride metabolic process
          negative regulation of neuron death
          baculum development
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          steroid binding
          protein binding
          beta-catenin binding
          transcription factor binding
          zinc ion binding
          enzyme binding
          estrogen receptor activity
          type 1 metabotropic glutamate receptor binding
          protein complex binding
          estrogen response element binding
          RNA polymerase II transcription factor activity, estrogen-activated sequence-specific DNA binding
          hormone binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2lcrw:80/100.66.78.62:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0