Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012733.4 | NP_036865.1 | 4 | 528 | ||
GenBank mRNA | FQ209770.1 | - | 4 | 542 | ||
FQ209927.1 | - | 4 | 512 | |||
FQ210835.1 | - | 4 | 543 | |||
FQ218090.1 | - | 4 | 542 | |||
M16459.1 | - | 4 | 528 | |||
M19257.1 | - | 4 | 459 |