Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APOA4 SIRNA Assays ›
Gene Symbol:
Apoa4
Gene Name:
apolipoprotein A-IV
Gene Aliases:
Apo-AIV, ApoA-IV, apoAIV
Chromosome Location:
Chr.8: 50536983 - 50539371 on Build Rnor_6.0
UniGene ID:
Rn.15739
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
200322
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Apoa4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012737.1
NP_036869.1
2
210
GenBank mRNA
BC091159.1
-
2
185
7373033
FQ218620.1
-
2
211
M00002.1
-
2
210
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_007468, M64249, BC050149, M64248, AK004856, and others.
Panther Classification:
Molecular Function -
apolipoprotein
Gene Ontology Categories:
Function(s)
Process(es)
innate immune response in mucosa
cholesterol biosynthetic process
lipid transport
response to lipid hydroperoxide
leukocyte cell-cell adhesion
cholesterol metabolic process
positive regulation of cholesterol esterification
positive regulation of triglyceride catabolic process
peripheral nervous system axon regeneration
removal of superoxide radicals
regulation of intestinal cholesterol absorption
response to food
regulation of cholesterol transport
cholesterol efflux
phospholipid efflux
response to triglyceride
very-low-density lipoprotein particle remodeling
high-density lipoprotein particle assembly
negative regulation of plasma lipoprotein particle oxidation
lipoprotein metabolic process
cholesterol homeostasis
hydrogen peroxide catabolic process
reverse cholesterol transport
multicellular organism lipid catabolic process
positive regulation of fatty acid biosynthetic process
phosphatidylcholine metabolic process
positive regulation of lipoprotein lipase activity
lipid homeostasis
protein-lipid complex assembly
triglyceride homeostasis
copper ion binding
lipid binding
cholesterol binding
antioxidant activity
cholesterol transporter activity
phosphatidylcholine binding
protein homodimerization activity
phosphatidylcholine-sterol O-acyltransferase activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0