Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADRA2A SIRNA Assays ›
Gene Symbol:
Adra2a
Gene Name:
adrenoceptor alpha 2A
Gene Aliases:
CA2-47, RATRG20, RG20
Chromosome Location:
Chr.1: 274766283 - 274769083 on Build Rnor_6.0
UniGene ID:
Rn.170171
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
200328
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Adra2a
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012739.3
NP_036871.3
1
629
GenBank mRNA
U79031.1
-
1
584
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_007417, AK046802, BC138531, AK140461, AK141573.
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cytokine production
acute inflammatory response
DNA replication
regulation of smooth muscle contraction
signal transduction
G-protein coupled receptor signaling pathway
female pregnancy
regulation of vasoconstriction
platelet activation
positive regulation of cell migration
negative regulation of cAMP biosynthetic process
activation of protein kinase activity
activation of protein kinase B activity
cellular response to hormone stimulus
epidermal growth factor-activated receptor transactivation by G-protein coupled receptor signaling pathway
glucose homeostasis
fear response
positive regulation of MAP kinase activity
positive regulation of epidermal growth factor-activated receptor activity
positive regulation of vasodilation
negative regulation of calcium ion-dependent exocytosis
negative regulation of insulin secretion
thermoception
negative regulation of lipid catabolic process
positive regulation of membrane protein ectodomain proteolysis
negative regulation of insulin secretion involved in cellular response to glucose stimulus
negative regulation of uterine smooth muscle contraction
adrenergic receptor signaling pathway
adenylate cyclase-inhibiting adrenergic receptor signaling pathway
phospholipase C-activating adrenergic receptor signaling pathway
activation of MAPK activity by adrenergic receptor signaling pathway
positive regulation of wound healing
adrenergic receptor activity
alpha2-adrenergic receptor activity
protein kinase binding
alpha-2C adrenergic receptor binding
thioesterase binding
heterotrimeric G-protein binding
protein homodimerization activity
protein heterodimerization activity
epinephrine binding
norepinephrine binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0