Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109882.1 | NP_001103352.1 | 3 | 561 | ||
NM_012783.3 | NP_036915.1 | 2 | 213 | |||
GenBank mRNA | AY120888.1 | - | 3 | 480 | ||
BC059145.1 | - | 2 | 137 | 6889785 | ||
CV119278.1 | - | 3 | 561 | |||
FQ212571.1 | - | 2 | 149 | |||
FQ212582.1 | - | 2 | 157 | |||
FQ213065.1 | - | 2 | 159 | |||
FQ213626.1 | - | 2 | 149 | |||
FQ214193.1 | - | 2 | 149 | |||
FQ214232.1 | - | 2 | 154 | |||
FQ219910.1 | - | 2 | 133 | |||
FQ219990.1 | - | 2 | 149 | |||
FQ220920.1 | - | 2 | 157 | |||
FQ224837.1 | - | 2 | 158 | |||
FQ229144.1 | - | 2 | 159 | |||
FQ229755.1 | - | 2 | 146 | |||
FQ230014.1 | - | 2 | 159 | |||
FQ231850.1 | - | 2 | 159 | |||
FQ232262.1 | - | 2 | 149 | |||
FQ234420.1 | - | 2 | 159 | |||
FQ234809.1 | - | 2 | 157 | |||
FQ234829.1 | - | 2 | 149 | |||
X54640.1 | - | 340 | ||||
X67215.1 | - | 2 | 134 |