Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2 SIRNA Assays ›
Gene Symbol:
Bcl2
Gene Name:
B-cell CLL/lymphoma 2
Gene Aliases:
Bcl-2
Chromosome Location:
Chr.13: 26605426 - 26769374 on Build Rnor_6.0
UniGene ID:
Rn.9996
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
200541
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Bcl2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_016993.1
NP_058689.1
2
987
GenBank mRNA
L14680.1
-
2
987
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_000633, M14745, BC027258, X06487, M13994.
This siRNA also targets the following transcripts in Mouse: NM_009741, AK049473, BC095964.
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
cell morphogenesis
ossification
ovarian follicle development
metanephros development
ureteric bud development
branching involved in ureteric bud morphogenesis
behavioral fear response
response to hypoxia
leukocyte homeostasis
B cell homeostasis
kidney development
release of cytochrome c from mitochondria
regulation of cell-matrix adhesion
lymphocyte homeostasis
lymphoid progenitor cell differentiation
B cell lineage commitment
T cell lineage commitment
immune system development
response to ischemia
renal system process
protein dephosphorylation
melanin metabolic process
regulation of nitrogen utilization
cellular calcium ion homeostasis
apoptotic process
cellular response to DNA damage stimulus
response to oxidative stress
actin filament organization
axonogenesis
brain development
aging
cell aging
response to nutrient
cell proliferation
positive regulation of cell proliferation
negative regulation of cell proliferation
male gonad development
extrinsic apoptotic signaling pathway via death domain receptors
intrinsic apoptotic signaling pathway in response to DNA damage
intrinsic apoptotic signaling pathway in response to oxidative stress
apoptotic mitochondrial changes
response to heat
response to toxic substance
post-embryonic development
organ morphogenesis
response to organic substance
response to inorganic substance
response to iron ion
response to aluminum ion
response to UV-B
response to gamma radiation
regulation of gene expression
negative regulation of calcium ion transport into cytosol
regulation of glycoprotein biosynthetic process
mesenchymal cell development
positive regulation of neuron maturation
response to organic cyclic compound
positive regulation of smooth muscle cell migration
cell growth
single organismal cell-cell adhesion
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
cochlear nucleus development
gland morphogenesis
regulation of transmembrane transporter activity
hemopoiesis
B cell differentiation
T cell differentiation
negative regulation of ossification
positive regulation of cell growth
negative regulation of cell growth
melanocyte differentiation
negative regulation of cell migration
positive regulation of B cell proliferation
response to caffeine
hair follicle morphogenesis
axon regeneration
regulation of protein stability
endoplasmic reticulum calcium ion homeostasis
glomerulus development
negative regulation of cellular pH reduction
response to insulin
regulation of protein localization
negative regulation of myeloid cell apoptotic process
T cell differentiation in thymus
positive regulation of peptidyl-serine phosphorylation
response to L-ascorbic acid
negative regulation of osteoblast proliferation
response to cytokine
glial cell apoptotic process
response to nicotine
organ growth
growth
positive regulation of multicellular organism growth
B cell proliferation
cellular response to glucose starvation
response to drug
response to hydrogen peroxide
regulation of apoptotic process
T cell homeostasis
negative regulation of apoptotic process
regulation of programmed cell death
positive regulation of catalytic activity
CD8-positive, alpha-beta T cell lineage commitment
response to peptide hormone
pigmentation
regulation of protein homodimerization activity
regulation of protein heterodimerization activity
negative regulation of neuron apoptotic process
ear development
response to estrogen
regulation of viral genome replication
response to ethanol
positive regulation of melanocyte differentiation
negative regulation of mitotic cell cycle
negative regulation of retinal cell programmed cell death
response to copper ion
regulation of mitochondrial membrane permeability
focal adhesion assembly
developmental pigmentation
regulation of developmental pigmentation
positive regulation of developmental pigmentation
spleen development
thymus development
response to steroid hormone
digestive tract morphogenesis
developmental growth
oocyte development
oligodendrocyte differentiation
positive regulation of skeletal muscle fiber development
pigment granule organization
homeostasis of number of cells within a tissue
regulation of catalytic activity
B cell receptor signaling pathway
response to glucocorticoid
neuron apoptotic process
response to corticosterone
response to cAMP
response to folic acid
response to electrical stimulus
defense response to virus
regulation of cell cycle
regulation of mitochondrial membrane potential
regulation of calcium ion transport
transmembrane transport
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
cellular response to organic substance
cellular response to hypoxia
reactive oxygen species metabolic process
extrinsic apoptotic signaling pathway in absence of ligand
negative regulation of G1/S transition of mitotic cell cycle
negative regulation of anoikis
negative regulation of apoptotic signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
negative regulation of intrinsic apoptotic signaling pathway
protease binding
protein binding
transcription factor binding
channel activity
channel inhibitor activity
protein phosphatase binding
ubiquitin protein ligase binding
identical protein binding
protein homodimerization activity
sequence-specific DNA binding
protein heterodimerization activity
BH domain binding
BH3 domain binding
protein phosphatase 2A binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0