Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ITGB1 SIRNA Assays ›
          Gene Symbol:
          Itgb1
          Gene Name:
          integrin, beta 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 61677512 - 61725537 on Build Rnor_6.0
          UniGene ID:
          Rn.25733
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 200621

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Itgb1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017022.2 NP_058718.2 7 1084
          XM_006255824.1 XP_006255886.1 7 1114
          GenBank mRNA BC131845.1 - 7 1084 8366766
          U12309.1 - 7 958

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          integrin

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          in utero embryonic development
          cell fate specification
          tissue homeostasis
          cell migration involved in sprouting angiogenesis
          cellular calcium ion homeostasis
          cell adhesion
          leukocyte cell-cell adhesion
          cell-matrix adhesion
          calcium-independent cell-matrix adhesion
          transforming growth factor beta receptor signaling pathway
          integrin-mediated signaling pathway
          regulation of G-protein coupled receptor protein signaling pathway
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          germ cell migration
          visual learning
          regulation of collagen catabolic process
          positive regulation of cell-substrate adhesion
          positive regulation of neuron projection development
          response to activity
          cell migration
          formation of radial glial scaffolds
          positive regulation of cell migration
          neuron projection development
          negative regulation of cell projection organization
          cell-substrate adhesion
          response to nutrient levels
          protein transport within lipid bilayer
          cell-cell adhesion mediated by integrin
          heterotypic cell-cell adhesion
          response to gonadotropin
          negative regulation of Rho protein signal transduction
          response to drug
          positive regulation of apoptotic process
          stress fiber assembly
          positive regulation of MAPK cascade
          sarcomere organization
          negative regulation of cell differentiation
          negative regulation of neuron differentiation
          positive regulation of neuron differentiation
          positive regulation of endocytosis
          mesodermal cell differentiation
          axon extension
          cardiac muscle tissue development
          dendrite morphogenesis
          positive regulation of peptidyl-tyrosine phosphorylation
          leukocyte tethering or rolling
          regulation of cell cycle
          cardiac muscle cell differentiation
          maternal process involved in female pregnancy
          bicellular tight junction assembly
          cellular response to mechanical stimulus
          cellular response to vitamin D
          cellular response to ionizing radiation
          response to transforming growth factor beta
          positive regulation of establishment of protein localization to plasma membrane
          negative regulation of anoikis
          glycoprotein binding
          fibronectin binding
          protease binding
          actin binding
          receptor activity
          receptor binding
          integrin binding
          protein binding
          collagen binding
          kinase binding
          protein kinase binding
          protein domain specific binding
          protein complex binding
          peptide binding
          laminin binding
          metal ion binding
          protein heterodimerization activity
          cell adhesion molecule binding
          alpha-actinin binding
          collagen binding involved in cell-matrix adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline