Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017041.1 | NP_058737.1 | 14 | 2047 | ||
XM_006233363.2 | XP_006233425.1 | 13 | 2514 | |||
XM_006233364.2 | XP_006233426.1 | 14 | 1809 | |||
GenBank mRNA | AF452728.1 | - | 1 | 117 | ||
D90035.1 | - | 14 | 2047 | |||
FQ212108.1 | - | 1 | 18 | |||
FQ212355.1 | - | 14 | 2391 | |||
FQ212693.1 | - | 1 | 18 | |||
FQ212817.1 | - | 9 | 1101 | |||
M29275.1 | - | 14 | 2047 | |||
X57115.1 | - | 13 | 2080 |