Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psg19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270654.1 | NP_001257583.1 | 2 | 419 | ||
NM_019126.2 | NP_061999.2 | 2 | 419 | |||
GenBank mRNA | BC097328.1 | - | 2 | 393 | 7462517 | |
U09815.1 | - | 419 |
Gene Symbol: Hand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC097327.1 | - | 391 | 7444552 |
Gene Symbol: LOC102557319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759035.1 | XP_008757257.1 | 2 | 392 |