Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM1 SIRNA Assays ›
Gene Symbol:
Tpm1
Gene Name:
tropomyosin 1, alpha
Gene Aliases:
Alpha-tm, Tma2, Tmsa
Chromosome Location:
Chr.8: 72814737 - 72841496 on Build Rnor_6.0
UniGene ID:
Rn.87540
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
200850
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034068.1
NP_001029240.1
4
509
NM_001034069.1
NP_001029241.1
4
509
NM_001034070.1
NP_001029242.1
4
509
NM_001034071.1
NP_001029243.1
4
509
NM_001034072.1
NP_001029244.1
4
509
NM_001034073.1
NP_001029245.1
3
446
NM_001034074.1
NP_001029246.1
3
446
NM_001034075.1
NP_001029247.1
3
446
NM_001301336.1
NP_001288265.1
4
509
NM_001301342.1
NP_001288271.1
4
509
NM_001301736.1
NP_001288665.1
3
446
NM_019131.2
NP_062004.1
3
446
XM_006243386.2
XP_006243448.1
4
501
XM_006243388.2
XP_006243450.1
4
501
XM_006243389.2
XP_006243451.1
4
501
XM_006243390.2
XP_006243452.1
4
501
XM_006243393.1
XP_006243455.1
3
444
XM_006243394.1
3
462
XM_006243395.2
XP_006243457.1
3
453
XM_008766207.1
XP_008764429.1
4
501
XM_008766208.1
XP_008764430.1
3
444
GenBank mRNA
AF372215.1
-
3
295
AF372216.1
-
4
408
AF372217.1
-
4
414
BC078780.1
-
3
413
7106337
BU670983.1
-
4
509
CB585296.1
-
3
449
CF108680.1
-
4
457
CK483405.1
-
4
463
FQ074026.1
-
4
567
FQ214619.1
-
4
484
FQ214803.1
-
4
480
FQ214809.1
-
4
478
FQ214835.1
-
4
478
FQ214864.1
-
4
482
FQ214868.1
-
4
478
FQ214869.1
-
4
484
FQ214872.1
-
4
477
FQ214877.1
-
4
478
FQ214894.1
-
4
484
FQ214902.1
-
4
478
FQ214911.1
-
4
480
FQ214944.1
-
4
484
FQ214946.1
-
4
484
FQ214991.1
-
4
478
FQ215107.1
-
4
478
FQ215186.1
-
4
484
FQ215188.1
-
4
478
FQ215206.1
-
4
513
FQ215226.1
-
4
484
FQ215459.1
-
4
480
FQ215512.1
-
4
484
FQ215522.1
-
4
555
FQ215524.1
-
4
480
FQ215538.1
-
4
484
FQ215698.1
-
4
478
FQ215703.1
-
4
478
FQ215724.1
-
4
565
FQ215746.1
-
4
480
FQ215761.1
-
4
478
FQ215803.1
-
4
484
FQ215808.1
-
4
480
FQ215815.1
-
4
478
FQ215856.1
-
4
478
FQ215919.1
-
4
486
FQ215944.1
-
4
485
FQ215969.1
-
4
478
FQ215995.1
-
4
480
FQ216014.1
-
4
562
FQ216018.1
-
4
484
FQ216052.1
-
4
478
FQ216082.1
-
4
562
FQ216105.1
-
4
478
FQ216118.1
-
4
562
FQ216145.1
-
4
484
FQ216160.1
-
4
477
FQ216319.1
-
4
480
FQ216340.1
-
4
484
FQ216368.1
-
4
480
FQ216400.1
-
4
480
FQ216494.1
-
4
478
FQ216552.1
-
4
484
FQ216557.1
-
4
484
FQ216615.1
-
4
393
FQ216637.1
-
4
480
FQ216683.1
-
4
480
FQ216697.1
-
4
480
FQ216747.1
-
4
480
FQ217468.1
-
3
229
FQ217503.1
-
4
484
FQ217616.1
-
2
46
FQ217637.1
-
4
484
FQ217808.1
-
4
480
FQ223706.1
-
4
480
FQ223993.1
-
4
482
FQ224036.1
-
4
395
FQ224207.1
-
4
478
FQ224838.1
-
4
484
FQ224943.1
-
4
482
FQ224961.1
-
4
480
M34134.1
-
3
431
M34135.1
-
4
443
M34136.1
-
2
161
M60666.1
-
4
456
M60667.1
-
4
456
M60668.1
-
3
393
M60669.1
-
3
393
X02411.1
-
4
407
X02412.1
-
2
163
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001164255, NM_001164254, NM_001164256, NM_001164250, NM_024427, and others.
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of heart rate by epinephrine
muscle contraction
muscle filament sliding
negative regulation of cell migration
ruffle organization
positive regulation of ATPase activity
cellular response to reactive oxygen species
wound healing
regulation of ATPase activity
sarcomere organization
positive regulation of cell adhesion
positive regulation of stress fiber assembly
actin filament capping
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
actin binding
cytoskeletal protein binding
protein homodimerization activity
protein N-terminus binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0