Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012334.2 | NP_036466.2 | 20, 21 | 2516 | ||
XM_005248306.4 | XP_005248363.1 | 2, 3 | 1410 | |||
XM_005248307.2 | XP_005248364.1 | 2, 3 | 886 | |||
XM_006714475.2 | XP_006714538.1 | 19, 20 | 2223 | |||
XM_011514046.2 | XP_011512348.1 | 2, 3 | 589 | |||
GenBank mRNA | AB018342.2 | - | 20, 21 | 2503 | ||
AF132021.1 | - | 20, 21 | 2128 | |||
AF234532.1 | - | 20, 21 | 2270 | |||
AF247457.2 | - | 20, 21 | 2201 | |||
AK024719.1 | - | 11, 12 | 1127 | |||
AK025921.1 | - | 1355 | ||||
AK125246.1 | - | 20, 21 | 2500 | |||
AK295736.1 | - | 2, 3 | 371 | |||
AK316138.1 | - | 2, 3 | 544 | |||
AL832428.1 | - | 10, 11 | 973 | |||
BC014956.1 | - | 20, 21 | 2512 | |||
BC137168.1 | - | 20, 21 | 2225 | 9021165 | ||
BC150285.1 | - | 20, 21 | 2503 | 8860110 |