Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270617.1 | NP_001257546.1 | 27 | 3398 | ||
NM_001270618.1 | NP_001257547.1 | 27 | 3398 | |||
XM_008762140.1 | XP_008760362.1 | 29 | 3662 | |||
XM_008762141.1 | XP_008760363.1 | 29 | 3661 | |||
XM_008762142.1 | XP_008760364.1 | 28 | 3566 | |||
XM_008762143.1 | XP_008760365.1 | 28 | 3566 | |||
XM_008762144.1 | XP_008760366.1 | 27 | 3470 | |||
GenBank mRNA | BC087017.1 | - | 27 | 3440 | 7109809 | |
EF577032.1 | - | 26 | 3382 | |||
JN579708.1 | - | 27 | 3398 | |||
U10096.1 | - | 27 | 3398 |