Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SERPINA1 SIRNA Assays ›
          Gene Symbol:
          Serpina1
          Gene Name:
          serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1
          Gene Aliases:
          AAT, Pi, Spi1
          Chromosome Location:
          Chr.6: 127610241 - 127632265 on Build Rnor_6.0
          UniGene ID:
          Rn.1419
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 200956

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Serpina1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022519.2 NP_071964.2 6 1419
          XM_006240456.1 XP_006240518.1 5 1276
          GenBank mRNA BC078824.1 - 6 1419 7131535
          D00675.1 - 5 1222
          FQ209479.1 - 4 733
          FQ209517.1 - 4 733
          FQ209677.1 - 4 733
          FQ209686.1 - 4 733
          FQ209716.1 - 4 734
          FQ209779.1 - 4 734
          FQ209814.1 - 4 734
          FQ209897.1 - 4 734
          FQ209914.1 - 4 733
          FQ210068.1 - 4 733
          FQ210655.1 - 5 1212
          FQ210668.1 - 3 355
          FQ210685.1 - 4 733
          FQ210775.1 - 4 733
          FQ210832.1 - 4 653
          FQ210989.1 - 4 734
          FQ210996.1 - 4 734
          FQ211012.1 - 4 733
          FQ211042.1 - 4 733
          FQ211203.1 - 4 733
          FQ211353.1 - 4 733
          FQ211455.1 - 4 599
          FQ218226.1 - 4 733
          FQ218241.1 - 4 733
          FQ218252.1 - 4 734
          FQ218261.1 - 4 733
          FQ218411.1 - 5 1352
          FQ218460.1 - 4 733
          FQ218555.1 - 4 733
          FQ218567.1 - 4 733
          FQ218586.1 - 4 733
          FQ218654.1 - 4 733
          FQ218694.1 - 4 734
          FQ218710.1 - 4 732
          FQ218857.1 - 4 734
          FQ218947.1 - 4 734
          FQ218955.1 - 4 734
          FQ218993.1 - 4 733
          FQ219224.1 - 4 733
          FQ219252.1 - 4 734
          FQ219268.1 - 4 733
          FQ219276.1 - 4 733
          FQ219303.1 - 4 733
          FQ219383.1 - 4 733
          FQ219441.1 - 4 733
          FQ219458.1 - 4 733
          FQ219621.1 - 4 733
          FQ219652.1 - 4 733
          FQ219732.1 - 4 733
          FQ219741.1 - 5 1213
          FQ219765.1 - 4 733
          M32247.1 - 4 1159

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          acute-phase response
          inflammatory response
          response to inorganic substance
          response to lead ion
          response to organic cyclic compound
          response to estradiol
          response to lipopolysaccharide
          response to methanol
          response to triglyceride
          response to cytokine
          response to chromate
          negative regulation of serine-type endopeptidase activity
          glycoprotein binding
          protease binding
          endopeptidase inhibitor activity
          serine-type endopeptidase inhibitor activity
          protein binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-cqs7j:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0