siRNA Details
Gene Symbol: Plp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030990.2 | NP_112252.1 | 3 | 549 | ||
XM_008773401.1 | XP_008771623.1 | 6 | 935 | |||
GenBank mRNA | BC093596.1 | - | 2 | 410 | 7314758 | |
FQ212680.1 | - | 2 | 263 | |||
FQ213540.1 | - | 2 | 263 | |||
FQ213548.1 | - | 2 | 263 | |||
FQ213841.1 | - | 2 | 263 | |||
FQ213943.1 | - | 2 | 263 | |||
FQ214012.1 | - | 2 | 263 | |||
FQ214050.1 | - | 2 | 263 | |||
FQ214153.1 | - | 2 | 263 | |||
M11185.1 | - | 3 | 524 | |||
M25888.1 | - | 1 | 89 | |||
X02809.1 | - | 3 | 453 |

More Information
Other Species Targeted

Panther Classification:
