Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYND8 SIRNA Assays ›
Gene Symbol:
ZMYND8
Gene Name:
zinc finger MYND-type containing 8
Gene Aliases:
PRKCBP1, PRO2893, RACK7
Chromosome Location:
Chr.20: 47209214 - 47357780 on Build GRCh38
UniGene ID:
Hs.446240
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
20109
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYND8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281771.2
NP_001268700.1
14
2273
NM_001281772.2
NP_001268701.1
16
2472
NM_001281773.2
NP_001268702.1
15
2419
NM_001281774.2
NP_001268703.1
15
2419
NM_001281775.2
NP_001268704.1
14
2348
NM_001281776.2
NP_001268705.1
14
2348
NM_001281777.2
NP_001268706.1
14
2273
NM_001281778.2
NP_001268707.1
14
2273
NM_001281779.2
NP_001268708.1
15
2479
NM_001281780.2
NP_001268709.1
15
2479
NM_001281781.2
NP_001268710.1
13
2132
NM_001281782.2
NP_001268711.1
14
2273
NM_001281783.2
NP_001268712.1
14
2348
NM_001281784.2
NP_001268713.1
13
2132
NM_012408.5
NP_036540.3
14
2348
NM_183047.3
NP_898868.1
14
2348
NM_183048.3
NP_898869.1
14
2273
XM_005260356.4
XP_005260413.1
14
2385
XM_005260358.2
XP_005260415.1
13
2127
XM_005260360.4
XP_005260417.1
14
2850
XM_005260362.2
XP_005260419.1
14
2125
XM_005260364.2
XP_005260421.1
14
2110
XM_005260366.2
XP_005260423.1
14
2141
XM_006723760.3
XP_006723823.1
14
2523
XM_006723762.3
XP_006723825.1
14
2224
XM_006723763.1
XP_006723826.1
14
2139
XM_006723766.1
XP_006723829.1
14
2083
XM_011528749.2
XP_011527051.1
14
3029
XM_011528751.1
XP_011527053.1
14
2133
XM_011528752.1
XP_011527054.1
12
1974
XM_011528753.1
XP_011527055.1
15
2144
XM_011528754.1
XP_011527056.1
14
2071
XM_017027760.1
XP_016883249.1
13
2130
XM_017027761.1
XP_016883250.1
14
2596
XM_017027762.1
XP_016883251.1
16
3363
XM_017027763.1
XP_016883252.1
14
2276
XM_017027764.1
XP_016883253.1
14
2795
XM_017027765.1
XP_016883254.1
14
2125
XM_017027766.1
XP_016883255.1
14
2125
XM_017027767.1
XP_016883256.1
14
2940
XM_017027768.1
XP_016883257.1
14
2200
XM_017027769.1
XP_016883258.1
14
2110
XM_017027770.1
XP_016883259.1
15
2144
XM_017027771.1
XP_016883260.1
14
2110
XM_017027772.1
XP_016883261.1
14
2083
XM_017027773.1
XP_016883262.1
14
2125
XM_017027774.1
XP_016883263.1
13
2103
XM_017027775.1
XP_016883264.1
14
2071
XM_017027776.1
XP_016883265.1
13
1969
XM_017027777.1
XP_016883266.1
13
2103
XM_017027778.1
XP_016883267.1
13
2115
GenBank mRNA
AB032951.1
-
14
2323
AF233453.1
-
3
549
AF273045.1
-
4
946
AF454056.1
-
14
2165
AK000275.1
-
14
2090
AK001175.1
-
1
286
AK026151.1
-
4
893
AK074262.1
-
4
883
AK122966.1
-
13
1969
AK225857.1
-
14
2156
AK294511.1
-
16
2282
AK295747.1
-
14
2232
AK299899.1
-
15
2231
AK309228.1
-
14
2142
AL137703.1
-
13
2084
BC030721.1
-
11
1848
BC136608.1
-
14
2104
9020598
BC144289.1
-
14
2104
9052809
BC146802.1
-
14
2114
8860316
BX641005.1
-
15
2364
DQ082998.1
-
13
2129
DQ082999.1
-
13
2129
DQ368669.1
-
13
2129
DQ368670.1
-
13
2129
DQ368671.1
-
13
2054
DQ368672.1
-
13
2054
DQ368673.1
-
13
2129
DQ368674.1
-
13
2054
DQ368675.1
-
14
2260
DQ368676.1
-
14
2260
DQ368677.1
-
12
1913
DQ368678.1
-
13
2054
DQ368679.1
-
13
2129
DQ368680.1
-
12
1913
U48251.1
-
1
309
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
positive regulation of filopodium assembly
positive regulation of dendritic spine development
modulation of excitatory postsynaptic potential
regulation of postsynaptic density protein 95 clustering
positive regulation of dendritic spine maintenance
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription corepressor activity
chromatin binding
transcription cofactor activity
protein binding
zinc ion binding
histone binding
protein N-terminus binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0