Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173094.2 | NP_775117.2 | 2 | 232 | ||
XM_006233002.2 | XP_006233064.1 | 2 | 261 | |||
GenBank mRNA | BC078695.1 | - | 2 | 232 | 7112644 | |
BC083543.1 | - | 2 | 232 | 7132009 | ||
M33648.1 | - | 2 | 250 |