Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mid1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206950.1 | NP_996833.1 | 2 | 1164 | ||
XM_006256672.2 | XP_006256734.1 | 2 | 984 | |||
GenBank mRNA | AB354580.1 | - | 2 | 1061 | ||
BC061715.1 | - | 2 | 1164 | 5598493 |