Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGIR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000960.3 | NP_000951.1 | 3 | 1905 | ||
XM_005259093.2 | XP_005259150.1 | 3 | 2635 | |||
XM_005259095.3 | XP_005259152.1 | 3 | 1888 | |||
GenBank mRNA | BC075814.1 | - | 3 | 1882 | 5223178 | |
BC110342.1 | - | 3 | 1868 | 5762329 | ||
D25418.1 | - | 3 | 1883 |
Set Membership: |
G-protein coupled receptor |