siRNA Details
Gene Symbol: KIR2DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014512.1 | NP_055327.1 | 7, 8 | 894 | ||
GenBank mRNA | AF022046.1 | - | 7, 8 | 894 | ||
AF285437.1 | - | 6, 7 | 841 | |||
AM999888.1 | - | 7, 8 | 889 | |||
X89892.1 | - | 7, 8 | 901 | |||
X98858.1 | - | 7, 8 | 881 |
Gene Symbol: KIR2DL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | Y13054.1 | - | 6, 7 | 767 |
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291696.1 | NP_001278625.1 | 6, 7 | 616 | ||
NM_001291700.1 | NP_001278629.1 | 5, 6 | 322 | |||
NM_001291701.1 | NP_001278630.1 | 6, 7 | 865 | |||
NM_012312.4 | NP_036444.1 | 7, 8 | 916 | |||
GenBank mRNA | AF074087.1 | - | 5, 6 | 278 | ||
AF285438.1 | - | 6, 7 | 841 | |||
AJ002102.1 | - | 7, 8 | 878 | |||
AJ002103.1 | - | 7, 8 | 881 | |||
AJ002104.1 | - | 6, 7 | 830 | |||
AJ002105.1 | - | 6, 7 | 830 | |||
AM409094.1 | - | 7, 8 | 882 | |||
AY366243.1 | - | 7, 8 | 891 | |||
AY366244.1 | - | 7, 8 | 880 | |||
BC119631.1 | - | 7, 8 | 900 | 40115568 | ||
BC119632.1 | - | 7, 8 | 900 | 40115570 | ||
BC126856.1 | - | 7, 8 | 883 | 40017301 | ||
L41347.1 | - | 7, 8 | 900 | |||
L76667.1 | - | 7, 8 | 598 | |||
L76668.1 | - | 6, 7 | 592 | |||
U24079.1 | - | 7, 8 | 879 | |||
X89893.1 | - | 7, 8 | 881 |
Gene Symbol: KIR2DS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281971.1 | NP_001268900.1 | 7, 8 | 948 | ||
NM_001281972.1 | NP_001268901.1 | 6, 7 | 897 | |||
NM_012314.5 | NP_036446.3 | 8, 9 | 970 | |||
GenBank mRNA | AF002255.1 | - | 8, 9 | 927 | ||
AJ417554.1 | - | 6, 7 | 839 | |||
AJ417555.1 | - | 6, 7 | 861 | |||
AY366245.1 | - | 7, 8 | 915 | |||
AY366246.1 | - | 7, 8 | 866 | |||
AY388472.1 | - | 7, 8 | 919 | |||
BC096694.1 | - | 7, 8 | 892 | 40026979 | ||
BC101977.1 | - | 7, 8 | 892 | 40004312 | ||
BC103693.1 | - | 7, 8 | 892 | 40004314 | ||
BC108918.1 | - | 8, 9 | 883 | 40017300 | ||
BC127084.1 | - | 7, 8 | 898 | 40120468 | ||
BC144566.1 | - | 7, 8 | 970 | 9053094 | ||
BC171788.1 | - | 7, 8 | 970 | 9054442 | ||
L76671.1 | - | 7, 8 | 892 | |||
U24077.1 | - | 7, 8 | 872 | |||
X94609.1 | - | 7, 8 | 936 |
Gene Symbol: KIR3DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF262970.1 | - | 6, 7 | 1085 |
Gene Symbol: KIR3DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083539.2 | NP_001077008.1 | 9, 10 | 1202 | ||
NM_001282170.1 | NP_001269099.1 | 8, 9 | 917 | |||
NM_001282171.1 | NP_001269100.1 | 7, 8 | 881 | |||
GenBank mRNA | AF022044.1 | - | 9, 10 | 1178 | ||
AK301140.1 | - | 9, 10 | 1202 | |||
BC098107.1 | - | 7, 8 | 855 | 40026977 | ||
BC098288.1 | - | 9, 10 | 1176 | 40026980 | ||
BC105679.1 | - | 9, 10 | 1176 | 40004313 | ||
EF370406.1 | - | 8, 9 | 1125 | |||
L76661.1 | - | 9, 10 | 1175 | |||
U73396.1 | - | 9, 10 | 1165 | |||
X97232.1 | - | 8, 9 | 880 | |||
X97233.1 | - | 9, 10 | 1165 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
