siRNA Details
Gene Symbol: KIR2DS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012314.5 | NP_036446.3 | 5 | 555 | ||
GenBank mRNA | AF002255.1 | - | 5 | 512 | ||
AF135564.1 | - | 4 | 307 | |||
AJ417555.1 | - | 3 | 446 | |||
AY366245.1 | - | 4 | 500 | |||
AY388472.1 | - | 4 | 504 | |||
BC096694.1 | - | 4 | 477 | 40026979 | ||
BC101977.1 | - | 4 | 477 | 40004312 | ||
BC103693.1 | - | 4 | 477 | 40004314 | ||
BC108918.1 | - | 5 | 468 | 40017300 | ||
BC125147.1 | - | 3 | 450 | |||
BC127084.1 | - | 4 | 483 | 40120468 | ||
BC144566.1 | - | 4 | 555 | 9053094 | ||
BC171788.1 | - | 4 | 555 | 9054442 | ||
L76671.1 | - | 4 | 477 | |||
U24077.1 | - | 4 | 457 | |||
U96188.1 | - | 3 | 403 | |||
X94609.1 | - | 4 | 521 |
Gene Symbol: KIR2DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014218.2 | NP_055033.2 | 4 | 504 | ||
XM_011526939.2 | XP_011525241.1 | 4 | 1109 | |||
XM_017026782.1 | XP_016882271.1 | 4 | 482 | |||
XM_017026783.1 | XP_016882272.1 | 4 | 1109 | |||
GenBank mRNA | AF022045.1 | - | 4 | 473 | ||
AF135560.1 | - | 2 | 403 | |||
AF285431.1 | - | 4 | 499 | |||
AF285432.1 | - | 3 | 426 | |||
AY366237.1 | - | 4 | 502 | |||
AY366238.1 | - | 4 | 448 | |||
AY366239.1 | - | 4 | 496 | |||
AY366240.1 | - | 4 | 504 | |||
AY366241.1 | - | 4 | 503 | |||
AY388471.1 | - | 4 | 496 | |||
AY789055.1 | - | 5 | 499 | |||
AY789056.1 | - | 4 | 466 | |||
BC069344.1 | - | 4 | 499 | 7262296 | ||
KJ699235.1 | - | 4 | 739 | |||
L41267.1 | - | 5 | 498 | |||
U24076.1 | - | 5 | 499 | |||
U24078.1 | - | 4 | 506 | |||
U96189.1 | - | 2 | 403 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 4 | 495 | ||
GenBank mRNA | AF285434.1 | - | 3 | 426 | ||
AY366242.1 | - | 4 | 495 | |||
KP260924.1 | - | 4 | 462 | |||
L76669.1 | - | 4 | 477 | |||
U24075.1 | - | 4 | 480 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015868.2 | NP_056952.2 | 4 | 503 | ||
GenBank mRNA | AF022048.1 | - | 4 | 466 | ||
AF135556.1 | - | 3 | 403 | |||
AF135567.1 | - | 2 | 292 | |||
AF285435.1 | - | 3 | 426 | |||
AY789057.1 | - | 4 | 466 | |||
BC032422.1 | - | 4 | 479 | 5220339 | ||
BC050730.1 | - | 4 | 497 | 5219336 | ||
BC110613.1 | - | 4 | 480 | 40031619 | ||
FN395076.1 | - | 4 | 466 | |||
FN552550.1 | - | 4 | 448 | |||
L41268.1 | - | 4 | 503 | |||
L76662.1 | - | 4 | 477 | |||
L76663.1 | - | 4 | 477 | |||
U24074.1 | - | 4 | 487 | |||
U73395.1 | - | 4 | 487 | |||
U96190.1 | - | 2 | 403 |
Gene Symbol: KIR2DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014512.1 | NP_055327.1 | 4 | 479 | ||
GenBank mRNA | AF022046.1 | - | 4 | 479 | ||
AF285437.1 | - | 3 | 426 | |||
AM999888.1 | - | 4 | 474 | |||
X89892.1 | - | 4 | 486 | |||
X98858.1 | - | 4 | 466 |
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 4 | 501 | ||
NM_001291696.1 | NP_001278625.1 | 3 | 201 | |||
NM_001291701.1 | NP_001278630.1 | 4 | 501 | |||
NM_012312.4 | NP_036444.1 | 4 | 501 | |||
GenBank mRNA | AF285438.1 | - | 3 | 426 | ||
AJ002102.1 | - | 4 | 463 | |||
AJ002103.1 | - | 4 | 466 | |||
AJ002104.1 | - | 4 | 466 | |||
AM409094.1 | - | 4 | 467 | |||
AY366243.1 | - | 4 | 476 | |||
AY366244.1 | - | 4 | 465 | |||
BC108917.1 | - | 4 | 468 | 40017299 | ||
BC119631.1 | - | 4 | 485 | 40115568 | ||
BC119632.1 | - | 4 | 485 | 40115570 | ||
BC125146.1 | - | 3 | 450 | |||
BC126856.1 | - | 4 | 468 | 40017301 | ||
FR750994.1 | - | 4 | 501 | |||
L41347.1 | - | 4 | 485 | |||
L76668.1 | - | 3 | 177 | |||
U24079.1 | - | 4 | 464 | |||
X89893.1 | - | 4 | 466 |
Gene Symbol: KIR2DS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012313.1 | NP_036445.1 | 4 | 477 | ||
GenBank mRNA | AF022047.1 | - | 4 | 470 | ||
AM851002.1 | - | 4 | 502 | |||
AY789063.2 | - | 3 | 450 | |||
L76670.1 | - | 4 | 477 | |||
X97231.1 | - | 4 | 525 |
Gene Symbol: KIR2DS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014513.2 | NP_055328.2 | 4 | 500 | ||
GenBank mRNA | AF208054.1 | - | 4 | 500 | ||
AF272389.1 | - | 5 | 450 | |||
AY366247.1 | - | 4 | 524 | |||
AY366248.1 | - | 5, 6 | 524 | |||
L76672.1 | - | 4 | 477 |
Gene Symbol: KIR3DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322168.1 | NP_001309097.1 | 5 | 814 | ||
NM_013289.2 | NP_037421.2 | 5 | 814 | |||
GenBank mRNA | AF022049.1 | - | 5 | 751 | ||
AF103010.1 | - | 5 | 751 | |||
AF103011.1 | - | 5 | 751 | |||
AF103012.1 | - | 5 | 751 | |||
AF103013.1 | - | 5 | 751 | |||
AF208688.1 | - | 3 | 688 | |||
AF208689.1 | - | 3 | 688 | |||
AF262968.1 | - | 3 | 670 | |||
AF262969.1 | - | 3 | 670 | |||
AF262970.1 | - | 3 | 670 | |||
AF262971.1 | - | 3 | 670 | |||
AF262972.1 | - | 3 | 670 | |||
AF262973.1 | - | 5 | 784 | |||
AF262974.1 | - | 5 | 784 | |||
AJ417556.1 | - | 3 | 700 | |||
AJ417557.1 | - | 3 | 700 | |||
AY366236.1 | - | 5 | 789 | |||
AY366249.1 | - | 5 | 751 | |||
AY366250.1 | - | 5 | 777 | |||
AY366251.1 | - | 5 | 751 | |||
AY366252.1 | - | 5 | 751 | |||
AY760050.1 | - | 5 | 751 | |||
AY789065.1 | - | 5 | 766 | |||
AY789066.1 | - | 5 | 751 | |||
AY789072.1 | - | 5 | 751 | |||
BC028206.1 | - | 5 | 778 | 5215674 | ||
BC101540.1 | - | 5 | 770 | 8069046 | ||
BC101542.1 | - | 5 | 770 | 8069048 | ||
DQ178659.1 | - | 5 | 751 | |||
EF462885.1 | - | 3 | 681 | |||
EF472674.1 | - | 3 | 681 | |||
EF472675.1 | - | 3 | 681 | |||
EF472676.1 | - | 3 | 681 | |||
EF472677.1 | - | 3 | 681 | |||
EF472678.1 | - | 3 | 681 | |||
EF472679.1 | - | 3 | 681 | |||
L41269.1 | - | 5 | 770 | |||
L76664.1 | - | 4 | 477 | |||
U30273.1 | - | 5 | 753 | |||
U30274.1 | - | 5 | 814 | |||
U31416.1 | - | 5 | 776 | |||
U33328.1 | - | 5 | 776 | |||
X94262.1 | - | 5 | 752 |
Gene Symbol: KIR3DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083539.2 | NP_001077008.1 | 5 | 788 | ||
NM_001282170.1 | NP_001269099.1 | 4 | 503 | |||
NM_001282171.1 | NP_001269100.1 | 3 | 467 | |||
GenBank mRNA | AF022044.1 | - | 5 | 764 | ||
AJ417558.3 | - | 3 | 700 | |||
AK301140.1 | - | 5 | 788 | |||
BC098107.1 | - | 3 | 441 | 40026977 | ||
BC098288.1 | - | 5 | 762 | 40026980 | ||
BC105679.1 | - | 5 | 762 | 40004313 | ||
EF370406.1 | - | 4 | 711 | |||
EF582383.1 | - | 5 | 751 | |||
L76661.1 | - | 5 | 762 | |||
U73396.1 | - | 5 | 751 | |||
X97232.1 | - | 4 | 466 | |||
X97233.1 | - | 5 | 751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
