Search
Search

ZNF215| Gene Symbol: ZNF215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013250.2 | NP_037382.2 | 7 | 1490 | ||
| XM_006718311.2 | XP_006718374.1 | 7 | 1530 | |||
| GenBank mRNA | AF056618.1 | - | 7 | 1490 | ||
| AK298365.1 | - | 5 | 1221 | |||
| AK302639.1 | - | 6 | 1400 | |||