Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013382.5 | NP_037514.2 | 3, 4 | 645 | ||
XM_011536675.2 | XP_011534977.1 | 3, 4 | 645 | |||
XM_011536676.2 | XP_011534978.1 | 2, 3 | 558 | |||
XM_011536677.2 | XP_011534979.1 | 3, 4 | 646 | |||
XM_017021244.1 | XP_016876733.1 | 3, 4 | 639 | |||
GenBank mRNA | AB209114.1 | - | 3, 4 | 467 | ||
AF105020.1 | - | 3, 4 | 648 | |||
AL554279.3 | - | 3, 4 | 509 | |||
AY090480.1 | - | 3, 4 | 648 | |||
BC031651.1 | - | 3, 4 | 535 | 5169145 | ||
BX248027.1 | - | 2, 3 | 538 | |||
CR627447.1 | - | 3, 4 | 215 |