Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NRG1 SIRNA Assays ›
          Gene Symbol:
          NRG1
          Gene Name:
          neuregulin 1
          Gene Aliases:
          ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, MSTP131, NDF, NRG1-IT2, SMDF
          Chromosome Location:
          Chr.8: 31639222 - 32771716 on Build GRCh38
          UniGene ID:
          Hs.453951
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 20476

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NRG1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001159995.2 NP_001153467.1 5 622
          NM_001159996.2 NP_001153468.1 2 351
          NM_001159999.2 NP_001153471.1 6 673
          NM_001160001.2 NP_001153473.1 4 571
          NM_001160002.1 NP_001153474.1 5 1029
          NM_001160004.2 NP_001153476.1 6 1080
          NM_001160005.1 NP_001153477.1 4 978
          NM_001160007.1 NP_001153479.1 4 978
          NM_001160008.1 NP_001153480.1 6 1080
          NM_001322197.1 NP_001309126.1 2 351
          NM_001322201.1 NP_001309130.1 5 622
          NM_001322202.1 NP_001309131.1 4 571
          NM_001322205.1 NP_001309134.1 2 1714
          NM_001322206.1 NP_001309135.1 2 1714
          NM_001322207.1 NP_001309136.1 2 1714
          NM_004495.3 NP_004486.2 6 1080
          NM_013956.4 NP_039250.2 6 1080
          NM_013957.4 NP_039251.2 6 1080
          NM_013958.3 NP_039252.2 6 1080
          NM_013959.3 NP_039253.1 2 1714
          NM_013960.4 NP_039254.1 6 1080
          NM_013962.2 NP_039256.2 4 1339
          NM_013964.4 NP_039258.1 6 1080
          XM_005273486.3 XP_005273543.1 2 1714
          XM_005273487.3 XP_005273544.1 2 1714
          XM_011544512.2 XP_011542814.1 6 863
          XM_017013365.1 XP_016868854.1 6 792
          XM_017013366.1 XP_016868855.1 6 792
          XM_017013367.1 XP_016868856.1 4 573
          XM_017013368.1 XP_016868857.1 4 910
          XM_017013369.1 XP_016868858.1 2 1682
          XM_017013370.1 XP_016868859.1 2 1677
          XM_017013371.1 XP_016868860.1 6 792
          XM_017013372.1 XP_016868861.1 6 792
          GenBank mRNA AB451331.1 - 6 563
          AB451475.1 - 6 563
          AF026146.1 - 6 1010
          AK097005.1 - 7 927
          AK223435.1 - 2 1221
          AK289927.1 - 2 1714
          AK290730.1 - 6 979
          AK293240.1 - 2 186
          AK293270.1 - 2 259
          AK298132.1 - 6 1080
          BC006492.2 - 6 1052
          BC007675.2 - 2 1371
          BC064587.1 - 2 1167
          BC073871.1 - 2 1335
          BC144446.1 - 6 628
          BC150609.1 - 6 628 9056979
          BT007389.1 - 2 728
          CN603652.1 - 6 591
          CN603654.1 - 4 489
          CN603661.1 - 2 351
          CR450288.1 - 2 728
          DC425692.1 - 2 271
          EF372273.1 - 4 407
          EF372274.1 - 4 407
          EF372275.1 - 5 458
          EF372276.1 - 5 458
          EF372277.1 - 4 407
          EF517296.1 - 6 509
          EF517297.1 - 5 458
          EU359055.1 - 4 458
          EU363508.1 - 5 509
          EU363509.1 - 4 458
          EU363510.1 - 6 560
          GQ983557.1 - 4 571
          GQ983558.1 - 4 398
          L12260.1 - 4 1367
          L12261.1 - 6 956
          L41827.1 - 2 1228
          M94165.1 - 6 695
          M94166.1 - 6 655
          M94167.1 - 6 1023
          M94168.1 - 6 1055
          S68256.1 - 6 635
          U02326.1 - 6 671
          U02327.1 - 4 199
          U02328.1 - 4 286
          U02329.1 - 4 210
          U02330.1 - 5 365

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001271122, NM_001271121, NM_031588, NM_001271119, NM_001271124, and others.
          This siRNA also targets the following transcripts in Mouse: NM_178591, AY227027, AK141037, BC151113, AY648975, and others.

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPK activity
          cell morphogenesis
          startle response
          cardiac conduction system development
          ventricular trabecula myocardium morphogenesis
          cell communication
          activation of transmembrane receptor protein tyrosine kinase activity
          nervous system development
          synapse assembly
          peripheral nervous system development
          locomotory behavior
          regulation of blood pressure
          positive regulation of cell proliferation
          embryo development
          neural crest cell development
          regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of phosphatidylinositol 3-kinase signaling
          glucose transport
          peptidyl-tyrosine phosphorylation
          glial cell fate commitment
          chemorepulsion involved in interneuron migration from the subpallium to the cortex
          mammary gland development
          positive regulation of myelination
          activation of protein kinase B activity
          intracellular signal transduction
          ERBB signaling pathway
          ERBB2 signaling pathway
          wound healing
          negative regulation of protein catabolic process
          myelination
          regulation of protein homodimerization activity
          regulation of protein heterodimerization activity
          positive regulation of GTPase activity
          neurotransmitter receptor metabolic process
          positive regulation of axon extension
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of Ras protein signal transduction
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          neuron fate commitment
          oligodendrocyte differentiation
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of secretion
          positive regulation of striated muscle cell differentiation
          positive regulation of protein kinase B signaling
          cardiac muscle cell differentiation
          cardiac muscle cell myoblast differentiation
          endocardial cell differentiation
          positive regulation of dendritic spine development
          positive regulation of calcineurin-NFAT signaling cascade
          positive regulation of protein targeting to mitochondrion
          positive regulation of protein localization to cell surface
          regulation of cell motility
          positive regulation of cardiac muscle cell differentiation
          negative regulation of neuron migration
          transcription cofactor activity
          protein tyrosine kinase activity
          Ras guanyl-nucleotide exchange factor activity
          cytokine activity
          ErbB-2 class receptor binding
          integrin binding
          growth factor activity
          transmembrane receptor protein tyrosine kinase activator activity
          receptor tyrosine kinase binding
          ErbB-3 class receptor binding
          chemorepellent activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-xmktn:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0