Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCR10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003684.1 | NP_001003684.1 | 2 | 490 | ||
NM_013387.3 | NP_037519.2 | 2 | 431 | |||
GenBank mRNA | AB028598.1 | - | 2 | 432 | ||
AF112217.1 | - | 2 | 420 | |||
AF161468.1 | - | 2 | 418 | |||
AF161500.1 | - | 2 | 488 | |||
AF161536.1 | - | 2 | 437 | |||
AK225973.1 | - | 2 | 416 | |||
BC005402.1 | - | 2 | 431 | 3961930 | ||
BC015971.1 | - | 2 | 425 | 4097106 | ||
BI754905.1 | - | 2 | 393 | |||
BU935406.1 | - | 490 | ||||
CB999839.1 | - | 2 | 433 | |||
CD674029.1 | - | 2 | 298 |