Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ST6GALNAC6 SIRNA Assays ›
          Gene Symbol:
          ST6GALNAC6
          Gene Name:
          ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 6
          Gene Aliases:
          SIAT7-F, SIAT7F, ST6GALNACVI
          Chromosome Location:
          Chr.9: 127885321 - 127906725 on Build GRCh38
          UniGene ID:
          Hs.109672
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 20559

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ST6GALNAC6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286999.1 NP_001273928.1 5, 6 887
          NM_001287000.1 NP_001273929.1 4, 5 832
          NM_001287001.1 NP_001273930.1 4, 5 857
          NM_001287002.1 NP_001273931.1 4, 5 828
          NM_013443.4 NP_038471.2 5, 6 887
          XM_011518610.2 XP_011516912.1 6, 7 1628
          XM_011518611.2 XP_011516913.1 5, 6 1573
          XM_017014658.1 XP_016870147.1 4, 5 1477
          XM_017014659.1 XP_016870148.1 6, 7 1628
          XM_017014660.1 XP_016870149.1 4, 5 1477
          XM_017014661.1 XP_016870150.1 5, 6 1573
          XM_017014662.1 XP_016870151.1 4, 5 807
          XM_017014663.1 XP_016870152.1 5, 6 967
          XM_017014664.1 XP_016870153.1 4, 5 857
          XM_017014665.1 XP_016870154.1 5, 6 894
          XM_017014666.1 XP_016870155.1 5, 6 967
          XM_017014667.1 XP_016870156.1 4, 5 1238
          GenBank mRNA AB035173.1 - 3, 4 685
          AJ507293.1 - 3, 4 624
          AK023900.1 - 4, 5 783
          AK025959.1 - 1, 2 226
          AK057100.1 - 4, 5 857
          AK226174.1 - 4, 5 713
          AY358672.1 - 4, 5 793
          BC006564.2 - 5, 6 804 3009935
          BC007802.2 - 5, 6 831 4128833
          BC016299.2 - 5, 6 831 3507812
          CR457318.1 - 3, 4 610
          KU178617.1 - 4, 5 712

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ganglioside biosynthetic process
          glycosylceramide metabolic process
          glycosphingolipid metabolic process
          glycoprotein metabolic process
          oligosaccharide metabolic process
          oligosaccharide biosynthetic process
          cell-cell recognition
          protein N-linked glycosylation via asparagine
          sialylation
          alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase activity
          sialyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0