Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271052.1 | NP_001257981.1 | 19 | 3493 | ||
NM_013447.3 | NP_038475.2 | 21 | 3952 | |||
GenBank mRNA | AK024426.1 | - | 11 | 2321 | ||
AK024454.1 | - | 6 | 1533 | |||
CR936819.1 | - | 15 | 2929 |
Set Membership: |
G-protein coupled receptor |