Search
Search

NOB1| Gene Symbol: NOB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014062.2 | NP_054781.1 | 2 | 234 | ||
| NR_074074.1 | 2 | 234 | ||||
| GenBank mRNA | AB026125.1 | - | 2 | 192 | ||
| AF190161.1 | - | 2 | 162 | |||
| AK001028.1 | - | 2 | 190 | |||
| AK314307.1 | - | 2 | 189 | |||
| AY487344.1 | - | 2 | 208 | |||
| BC064630.1 | - | 2 | 177 | 5740890 | ||
| BG386829.1 | - | 1 | 178 | |||
| BM827373.1 | - | 2 | 210 | |||
| DA004856.1 | - | 2 | 190 | |||
| DB048531.1 | - | 2 | 234 | |||