Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNRHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000406.2 | NP_000397.1 | 1, 2 | 2273 | ||
GenBank mRNA | AY392011.1 | - | 1, 2 | 522 | ||
BC113546.1 | - | 1, 2 | 548 | 8322598 | ||
L03380.1 | - | 1, 2 | 577 | |||
L07949.1 | - | 1, 2 | 546 | |||
S60587.1 | - | 1, 2 | 546 | |||
S77472.1 | - | 1, 2 | 547 |
Set Membership: |
G-protein coupled receptor |