Search
Search

HCST| Gene Symbol: HCST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007469.1 | NP_001007470.1 | 2 | 189 | ||
| NM_014266.3 | NP_055081.1 | 2 | 189 | |||
| XM_017026193.1 | XP_016881682.1 | 2 | 304 | |||
| GenBank mRNA | AF072844.1 | - | 2 | 131 | ||
| AF122904.1 | - | 2 | 131 | |||
| AF172929.1 | - | 2 | 166 | |||
| AF285447.1 | - | 2 | 106 | |||
| AL050163.1 | - | 2 | 189 | |||
| AY359058.1 | - | 2 | 130 | |||
| BC035931.1 | - | 2 | 140 | 4701500 | ||
| BC046348.1 | - | 2 | 189 | 5229680 | ||
| BC065224.1 | - | 2 | 145 | 5762412 | ||