Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RBFOX2 SIRNA Assays ›
          Gene Symbol:
          RBFOX2
          Gene Name:
          RNA binding protein, fox-1 homolog 2
          Gene Aliases:
          FOX2, Fox-2, HNRBP2, HRNBP2, RBM9, RTA, dJ106I20.3, fxh
          Chromosome Location:
          Chr.22: 35738736 - 36028892 on Build GRCh38
          UniGene ID:
          Hs.282998
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 21017

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RBFOX2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001031695.2 NP_001026865.1 4, 5 823
          NM_001082576.1 NP_001076045.1 4, 5 823
          NM_001082577.1 NP_001076046.1 4, 5 823
          NM_001082578.1 NP_001076047.1 5, 6 783
          NM_001082579.1 NP_001076048.1 5, 6 780
          NM_014309.2 NP_055124.1 4, 5 826
          XM_005261428.2 XP_005261485.1 5, 6 1138
          XM_005261429.2 XP_005261486.1 5, 6 1135
          XM_005261430.2 XP_005261487.1 5, 6 1138
          XM_005261431.2 XP_005261488.1 5, 6 1138
          XM_005261432.2 XP_005261489.1 5, 6 1135
          XM_005261433.2 XP_005261490.1 5, 6 1138
          XM_005261435.1 XP_005261492.1 4, 5 827
          XM_005261437.1 XP_005261494.1 4, 5 830
          XM_006724185.2 XP_006724248.1 5, 6 1138
          XM_006724186.2 XP_006724249.1 5, 6 1135
          XM_006724187.2 XP_006724250.1 5, 6 1138
          XM_006724188.2 XP_006724251.1 5, 6 1138
          XM_006724189.3 XP_006724252.1 5, 6 831
          XM_006724190.1 XP_006724253.1 5, 6 525
          XM_006724191.1 XP_006724254.1 4, 5 830
          XM_006724192.2 XP_006724255.1 4, 5 660
          XM_006724193.3 XP_006724256.1 5, 6 1138
          XM_006724194.2 XP_006724257.1 4, 5 602
          XM_017028686.1 XP_016884175.1 5, 6 517
          XM_017028687.1 XP_016884176.1 5, 6 698
          XM_017028688.1 XP_016884177.1 5, 6 1135
          XM_017028689.1 XP_016884178.1 5, 6 603
          XM_017028690.1 XP_016884179.1 4, 5 580
          XM_017028691.1 XP_016884180.1 4, 5 577
          XM_017028692.1 XP_016884181.1 5, 6 872
          XM_017028693.1 XP_016884182.1 4, 5 830
          XM_017028694.1 XP_016884183.1 4, 5 827
          XM_017028695.1 XP_016884184.1 5, 6 587
          XM_017028696.1 XP_016884185.1 4, 5 830
          XM_017028697.1 XP_016884186.1 5, 6 1607
          XM_017028698.1 XP_016884187.1 4, 5 827
          GenBank mRNA AB649123.1 - 4, 5 458
          AF229058.1 - 4, 5 628
          AK055213.1 - 5, 6 744
          AK291460.1 - 4, 5 707
          AK314632.1 - 4, 5 642
          AL009266.1 - 4, 5 631
          AM419009.1 - 4, 5 482
          AY072786.1 - 5, 6 593
          AY960684.1 - 4, 5 458
          BC025281.1 - 4, 5 646 4555865
          CR456559.1 - 4, 5 524
          DQ778625.1 - 5, 6 671

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001079895, XM_006241845, XM_006241837, XM_006241849, XM_008765586, and others.
          This siRNA also targets the following transcripts in Mouse: NM_053104, NM_001110828, NM_001110830, NM_001110827, NM_175387, and others.

          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          mRNA processing
          nervous system development
          RNA splicing
          fibroblast growth factor receptor signaling pathway
          regulation of definitive erythrocyte differentiation
          RNA metabolic process
          radial glia guided migration of Purkinje cell
          intracellular estrogen receptor signaling pathway
          regulation of cell proliferation
          negative regulation of transcription, DNA-templated
          dendrite morphogenesis
          neuromuscular process controlling balance
          nucleotide binding
          transcription corepressor activity
          RNA binding
          mRNA binding
          protein binding
          transcription factor binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0