Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014228.4 | NP_055043.2 | 6, 7 | 1333 | ||
XM_017009767.1 | XP_016865256.1 | 7, 8 | 1240 | |||
XM_017009768.1 | XP_016865257.1 | 7, 8 | 1316 | |||
XM_017009769.1 | XP_016865258.1 | 7, 8 | 2847 | |||
XM_017009770.1 | XP_016865259.1 | 7, 8 | 967 | |||
GenBank mRNA | AK096607.1 | - | 6, 7 | 1237 | ||
AK308895.1 | - | 6, 7 | 1109 | |||
AK312309.1 | - | 6, 7 | 1113 | |||
BC069631.1 | - | 6, 7 | 866 | 7262437 | ||
BC093785.1 | - | 6, 7 | 942 | 7939630 | ||
BC113425.1 | - | 6, 7 | 942 | 8322477 | ||
S80071.1 | - | 6, 7 | 866 |