Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RBFOX2 SIRNA Assays ›
          Gene Symbol:
          RBFOX2
          Gene Name:
          RNA binding protein, fox-1 homolog 2
          Gene Aliases:
          FOX2, Fox-2, HNRBP2, HRNBP2, RBM9, RTA, dJ106I20.3, fxh
          Chromosome Location:
          Chr.22: 35738736 - 36028892 on Build GRCh38
          UniGene ID:
          Hs.282998
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 21107

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RBFOX2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001031695.2 NP_001026865.1 5, 6 916
          NM_001082576.1 NP_001076045.1 5, 6 916
          NM_001082577.1 NP_001076046.1 5, 6 916
          NM_001082578.1 NP_001076047.1 6, 7 876
          NM_001082579.1 NP_001076048.1 6, 7 873
          NM_014309.2 NP_055124.1 5, 6 919
          XM_005261428.2 XP_005261485.1 6, 7 1231
          XM_005261429.2 XP_005261486.1 6, 7 1228
          XM_005261430.2 XP_005261487.1 6, 7 1231
          XM_005261431.2 XP_005261488.1 6, 7 1231
          XM_005261432.2 XP_005261489.1 6, 7 1228
          XM_005261433.2 XP_005261490.1 6, 7 1231
          XM_005261435.1 XP_005261492.1 5, 6 920
          XM_005261437.1 XP_005261494.1 5, 6 923
          XM_006724185.2 XP_006724248.1 6, 7 1231
          XM_006724186.2 XP_006724249.1 6, 7 1228
          XM_006724187.2 XP_006724250.1 6, 7 1231
          XM_006724188.2 XP_006724251.1 6, 7 1231
          XM_006724189.3 XP_006724252.1 6, 7 924
          XM_006724190.1 XP_006724253.1 6, 7 618
          XM_006724191.1 XP_006724254.1 5, 6 923
          XM_006724192.2 XP_006724255.1 5, 6 753
          XM_006724193.3 XP_006724256.1 6, 7 1231
          XM_006724194.2 XP_006724257.1 5, 6 695
          XM_017028686.1 XP_016884175.1 6, 7 610
          XM_017028687.1 XP_016884176.1 6, 7 791
          XM_017028688.1 XP_016884177.1 6, 7 1228
          XM_017028689.1 XP_016884178.1 6, 7 696
          XM_017028690.1 XP_016884179.1 5, 6 673
          XM_017028691.1 XP_016884180.1 5, 6 670
          XM_017028692.1 XP_016884181.1 6, 7 965
          XM_017028693.1 XP_016884182.1 5, 6 923
          XM_017028694.1 XP_016884183.1 5, 6 920
          XM_017028695.1 XP_016884184.1 6, 7 680
          XM_017028696.1 XP_016884185.1 5, 6 923
          XM_017028697.1 XP_016884186.1 6, 7 1700
          XM_017028698.1 XP_016884187.1 5, 6 920
          GenBank mRNA AB649123.1 - 5, 6 551
          AF229058.1 - 5, 6 721
          AK055213.1 - 6, 7 837
          AK291460.1 - 5, 6 800
          AK314632.1 - 5, 6 735
          AL009266.1 - 5, 6 724
          AM419009.1 - 5, 6 575
          AY072786.1 - 6, 7 686
          AY960684.1 - 5, 6 551
          BC013115.1 - 4, 5 686 4346741
          BC025281.1 - 5, 6 739 4555865
          CR456559.1 - 5, 6 617
          DQ778625.1 - 6, 7 764

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          mRNA processing
          nervous system development
          RNA splicing
          fibroblast growth factor receptor signaling pathway
          regulation of definitive erythrocyte differentiation
          RNA metabolic process
          radial glia guided migration of Purkinje cell
          intracellular estrogen receptor signaling pathway
          regulation of cell proliferation
          negative regulation of transcription, DNA-templated
          dendrite morphogenesis
          neuromuscular process controlling balance
          nucleotide binding
          transcription corepressor activity
          RNA binding
          mRNA binding
          protein binding
          transcription factor binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-sskjb:80/100.66.75.127:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0