Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPSAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003294.3 | NP_003285.2 | 6 | 798 | ||
GenBank mRNA | AF206665.1 | - | 5 | 756 | ||
AF206666.1 | - | 4 | 674 | |||
AF206667.1 | - | 4 | 560 | |||
BC038416.1 | - | 5 | 752 | |||
BC051852.1 | - | 5 | 751 | |||
BC065923.1 | - | 5 | 957 | 5752632 | ||
BC074974.2 | - | 6 | 779 | 30915512 | ||
BC074975.2 | - | 6 | 779 | 30915294 | ||
CR541836.1 | - | 5 | 746 | |||
FJ931118.1 | - | 4 | 595 | |||
M30038.1 | - | 5 | 763 | |||
M33491.1 | - | 5 | 742 | |||
M37488.1 | - | 5 | 751 |
Gene Symbol: TPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024164.5 | NP_077078.5 | 6 | 769 | ||
GenBank mRNA | AK308208.1 | - | 6 | 770 | ||
BC028059.1 | - | 5 | 758 | 5181815 | ||
BC029356.1 | - | 6 | 769 | 4695458 | ||
FJ931119.1 | - | 4 | 600 | |||
FJ931120.1 | - | 4 | 599 | |||
M33492.1 | - | 5 | 744 | |||
M33493.1 | - | 5 | 723 | |||
S55551.1 | - | 5 | 753 |