siRNA Details
Gene Symbol: KIR2DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014512.1 | NP_055327.1 | 1, 2 | 41 | ||
GenBank mRNA | AF022046.1 | - | 1, 2 | 41 | ||
AM999888.1 | - | 1, 2 | 36 | |||
X89892.1 | - | 1, 2 | 48 | |||
X98858.1 | - | 1, 2 | 28 |
Gene Symbol: 1060P11.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ630586.1 | - | 1, 2 | 29 |
Gene Symbol: KIR2DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014218.2 | NP_055033.2 | 1, 2 | 66 | ||
XM_011526939.2 | XP_011525241.1 | 1, 2 | 671 | |||
XM_017026783.1 | XP_016882272.1 | 1, 2 | 671 | |||
GenBank mRNA | AF022045.1 | - | 1, 2 | 35 | ||
AF285431.1 | - | 1, 2 | 61 | |||
AY366237.1 | - | 1, 2 | 64 | |||
AY366239.1 | - | 1, 2 | 58 | |||
AY366240.1 | - | 1, 2 | 66 | |||
AY366241.1 | - | 1, 2 | 65 | |||
AY388471.1 | - | 1, 2 | 58 | |||
AY789055.1 | - | 1, 2 | 61 | |||
AY789056.1 | - | 1, 2 | 28 | |||
BC069344.1 | - | 1, 2 | 61 | 7262296 | ||
KJ699235.1 | - | 1, 2 | 301 | |||
L41267.1 | - | 1, 2 | 60 | |||
U24076.1 | - | 1, 2 | 61 | |||
U24078.1 | - | 1, 2 | 68 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 1, 2 | 57 | ||
GenBank mRNA | AY366242.1 | - | 1, 2 | 57 | ||
KP260924.1 | - | 1, 2 | 24 | |||
L76669.1 | - | 1, 2 | 39 | |||
U24075.1 | - | 1, 2 | 42 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF022048.1 | - | 1, 2 | 28 | ||
U73395.1 | - | 1, 2 | 49 |
Gene Symbol: KIR2DL5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020535.3 | NP_065396.1 | 1, 2 | 83 | ||
GenBank mRNA | AF204903.1 | - | 1, 2 | 83 | ||
AF217487.1 | - | 1, 2 | 52 |
Gene Symbol: KIR2DL5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018081.1 | NP_001018091.1 | 1, 2 | 28 | ||
GenBank mRNA | AF271607.1 | - | 1, 2 | 28 | ||
AF271608.1 | - | 1, 2 | 28 | |||
AJ544587.1 | - | 1, 2 | 29 |
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 1, 2 | 63 | ||
NM_001291696.1 | NP_001278625.1 | 1, 2 | 63 | |||
NM_001291700.1 | NP_001278629.1 | 1, 2 | 63 | |||
NM_001291701.1 | NP_001278630.1 | 1, 2 | 63 | |||
NM_012312.4 | NP_036444.1 | 1, 2 | 63 | |||
GenBank mRNA | AF074087.1 | - | 1, 2 | 19 | ||
AJ002103.1 | - | 1, 2 | 28 | |||
AJ002104.1 | - | 1, 2 | 28 | |||
AJ002105.1 | - | 1, 2 | 28 | |||
AM409094.1 | - | 1, 2 | 29 | |||
AY366243.1 | - | 1, 2 | 38 | |||
AY366244.1 | - | 1, 2 | 27 | |||
BC108917.1 | - | 1, 2 | 30 | 40017299 | ||
BC119631.1 | - | 1, 2 | 47 | 40115568 | ||
BC119632.1 | - | 1, 2 | 47 | 40115570 | ||
BC126856.1 | - | 1, 2 | 30 | 40017301 | ||
FR750994.1 | - | 1, 2 | 63 | |||
L41347.1 | - | 1, 2 | 47 | |||
L76667.1 | - | 1, 2 | 39 | |||
L76668.1 | - | 1, 2 | 39 | |||
U24079.1 | - | 1, 2 | 26 | |||
X89893.1 | - | 1, 2 | 28 |
Gene Symbol: KIR2DS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC108918.1 | - | 1, 2 | 30 | 40017300 |
Gene Symbol: KIR3DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322168.1 | NP_001309097.1 | 1, 2 | 91 | ||
NM_013289.2 | NP_037421.2 | 1, 2 | 91 | |||
GenBank mRNA | AF022049.1 | - | 1, 2 | 28 | ||
AF103010.1 | - | 1, 2 | 28 | |||
AF103011.1 | - | 1, 2 | 28 | |||
AF103012.1 | - | 1, 2 | 28 | |||
AF103013.1 | - | 1, 2 | 28 | |||
AF262973.1 | - | 1, 2 | 61 | |||
AF262974.1 | - | 1, 2 | 61 | |||
AY366236.1 | - | 1, 2 | 66 | |||
AY366249.1 | - | 1, 2 | 28 | |||
AY366250.1 | - | 1, 2 | 54 | |||
AY366251.1 | - | 1, 2 | 28 | |||
AY366252.1 | - | 1, 2 | 28 | |||
AY760050.1 | - | 1, 2 | 28 | |||
AY789065.1 | - | 1, 2 | 43 | |||
AY789066.1 | - | 1, 2 | 28 | |||
AY789072.1 | - | 1, 2 | 28 | |||
BC028206.1 | - | 1, 2 | 55 | 5215674 | ||
BC101540.1 | - | 1, 2 | 47 | 8069046 | ||
BC101542.1 | - | 1, 2 | 47 | 8069048 | ||
DQ178659.1 | - | 1, 2 | 28 | |||
L41269.1 | - | 1, 2 | 47 | |||
L76664.1 | - | 1, 2 | 39 | |||
U30273.1 | - | 1, 2 | 30 | |||
U30274.1 | - | 1, 2 | 91 | |||
U31416.1 | - | 1, 2 | 53 | |||
U33328.1 | - | 1, 2 | 53 | |||
X94262.1 | - | 1, 2 | 29 |
Gene Symbol: KIR3DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153443.4 | NP_703144.3 | 1, 2 | 78 | ||
GenBank mRNA | AF352324.1 | - | 1, 2 | 46 | ||
AJ938052.1 | - | 1, 2 | 33 | |||
AJ938053.1 | - | 1, 2 | 33 | |||
AJ938054.1 | - | 1, 2 | 33 | |||
AJ938055.1 | - | 1, 2 | 33 | |||
AJ938056.1 | - | 1, 2 | 33 | |||
AJ938057.1 | - | 1, 2 | 33 | |||
AJ938058.1 | - | 1, 2 | 33 | |||
AJ938059.1 | - | 1, 2 | 33 | |||
AJ938060.1 | - | 1, 2 | 33 | |||
AJ938061.1 | - | 1, 2 | 33 | |||
AJ938062.1 | - | 1, 2 | 33 | |||
AJ938063.1 | - | 1, 2 | 33 | |||
AJ938064.1 | - | 1, 2 | 33 | |||
AJ938065.1 | - | 1, 2 | 33 | |||
AJ938066.1 | - | 1, 2 | 33 | |||
BC136283.1 | - | 1, 2 | 78 | 9020270 | ||
BC143802.1 | - | 1, 2 | 78 | 9052316 |
Gene Symbol: KIR3DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083539.2 | NP_001077008.1 | 1, 2 | 65 | ||
NM_001282170.1 | NP_001269099.1 | 1, 2 | 65 | |||
GenBank mRNA | AF022044.1 | - | 1, 2 | 41 | ||
AK301140.1 | - | 1, 2 | 65 | |||
BC098288.1 | - | 1, 2 | 39 | 40026980 | ||
BC105679.1 | - | 1, 2 | 39 | 40004313 | ||
DC390157.1 | - | 1, 2 | 65 | |||
EF582383.1 | - | 1, 2 | 28 | |||
L76661.1 | - | 1, 2 | 39 | |||
U73396.1 | - | 1, 2 | 28 | |||
X97232.1 | - | 1, 2 | 28 | |||
X97233.1 | - | 1, 2 | 28 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
